  • Search Results
  • Saururus chinensis Baill induces apoptosis through endoplasmic reticulum stress in HepG2 hepatocellular carcinoma cells.
Saururus chinensis Baill induces apoptosis through endoplasmic reticulum stress in HepG2 hepatocellular carcinoma cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2015-06-28)
Ah Young Lee, Young-Ah Han, Ji-Eun Kim, Seong-Ho Hong, Eun-Jung Park, Myung-Haing Cho
ABSTRACT
In this study, we examined the mechanism underlying the effect of Saururus chinensis Baill (saururaceae) on hepatocellular carcinoma HepG2 cells. HepG2 cells and Chang cells were exposed to various concentrations of S. chinensis Baill extract (SC-E) for 24 h. SC-E affected more significantly HepG2 cells than Chang cells in terms of cell viability and ATP production. Therefore, current study examined detailed mechanism how SC-E affected HepG2 cell survival. We found that SC-E (75 and 150 μg/ml) induced apoptosis via oxidative stress. SC-E also caused CCAAT-enhancer-binding protein homologous protein (CHOP) activation by dissociating the binding immunoglobulin protein (BiP) from inositol-requiring 1α (IRE1α) in the endoplasmic reticulum (ER) and induced Bax, cytochrome c release to cytosol, caspase-3 activation, and poly ADP ribose polymerase (PARP) cleavage, resulting in HepG2 cell apoptosis. Furthermore, SC-E caused ER Ca(2+) leakage into the cytosol; ER dilation and mitochondrial membrane damage were observed in transmission electron microscopy (TEM). Taken together, our results demonstrated that SC-E induced cancer cell apoptosis specifically through ER stress.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
Sodium phenylbutyrate, ≥98% (HPLC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications