  • Search Results
  • Electrochemical impedance immunosensor for rapid detection of stressed pathogenic Staphylococcus aureus bacteria.
Electrochemical impedance immunosensor for rapid detection of stressed pathogenic Staphylococcus aureus bacteria.
Environmental science and pollution research international (2015-06-04)
Karima Bekir, Houcine Barhoumi, Mohamed Braiek, Amani Chrouda, Nadia Zine, Nabil Abid, Abdelrazek Maaref, Amina Bakhrouf, Hafedh Ben Ouada, Nicole Jaffrezic-Renault, Hedi Ben Mansour
ABSTRACT
In this work, we report the adaptation of bacteria to stress conditions that induce instability of their cultural, morphological, and enzymatic characters, on which the identification of pathogenic bacteria is based. These can raise serious issues during the characterization of bacteria. The timely detection of pathogens is also a subject of great importance. For this reason, our objective is oriented towards developing an immunosensing system for rapid detection and quantification of Staphylococcus aureus. Polyclonal anti-S. aureus are immobilized onto modified gold electrode by self-assembled molecular monolayer (SAM) method. The electrochemical performances of the developed immunosensor were evaluated by impedance spectroscopy through the monitoring of the charge transfer resistance at the modified solid/liquid interface using ferri-/ferrocyanide as redox probe. The developed immunosensor was applied to detect stressed and resuscitate bacteria. As a result, a stable and reproducible immunosensor with sensitivity of 15 kΩ/decade and a detection limit of 10 CFU/mL was obtained for the S. aureus concentrations ranging from 10(1) to 10(7) CFU/mL. A low deviation in the immunosensor response (±10 %) was signed when it is exposed to stressed and not stressed bacteria.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)