  • Search Results
  • The role of GTF2IRD1 in the auditory pathology of Williams-Beuren Syndrome.
The role of GTF2IRD1 in the auditory pathology of Williams-Beuren Syndrome.
European journal of human genetics : EJHG (2014-09-25)
Cesar P Canales, Ann C Y Wong, Peter W Gunning, Gary D Housley, Edna C Hardeman, Stephen J Palmer
ABSTRACT
Williams-Beuren Syndrome (WBS) is a rare genetic condition caused by a hemizygous deletion involving up to 28 genes within chromosome 7q11.23. Among the spectrum of physical and neurological defects in WBS, it is common to find a distinctive response to sound stimuli that includes extreme adverse reactions to loud, or sudden sounds and a fascination with certain sounds that may manifest as strengths in musical ability. However, hearing tests indicate that sensorineural hearing loss (SNHL) is frequently found in WBS patients. The functional and genetic basis of this unusual auditory phenotype is currently unknown. Here, we investigated the potential involvement of GTF2IRD1, a transcription factor encoded by a gene located within the WBS deletion that has been implicated as a contributor to the WBS assorted neurocognitive profile and craniofacial abnormalities. Using Gtf2ird1 knockout mice, we have analysed the expression of the gene in the inner ear and examined hearing capacity by evaluating the auditory brainstem response (ABR) and the distortion product of otoacoustic emissions (DPOAE). Our results show that Gtf2ird1 is expressed in a number of cell types within the cochlea, and Gtf2ird1 null mice showed higher auditory thresholds (hypoacusis) in both ABR and DPOAE hearing assessments. These data indicate that the principal hearing deficit in the mice can be traced to impairments in the amplification process mediated by the outer hair cells and suggests that similar mechanisms may underpin the SNHL experienced by WBS patients.
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution