  • Search Results
  • Synergistic fungicidal activity of the lipopeptide bacillomycin D with amphotericin B against pathogenic Candida species.
Synergistic fungicidal activity of the lipopeptide bacillomycin D with amphotericin B against pathogenic Candida species.
FEMS yeast research (2015-05-10)
Olfa Tabbene, Antonio Di Grazia, Sana Azaiez, Imen Ben Slimene, Salem Elkahoui, Mohamed Najib Alfeddy, Bruno Casciaro, Vincenzo Luca, Ferid Limam, Maria Luisa Mangoni
ABSTRACT
In the present study, the synergism of the lipopeptide bacillomycin D in combination with the polyene amphotericin B against pathogenic Candida species is described along with their potential cytotoxicity against mammalian cells. Bacillomycin D inhibited the growth of various Candida species at minimal concentrations from 12.5 to 25 μg ml(-1). Furthermore, it showed a synergistic effect with the antifungal drug amphotericin B in inhibiting the growth of Candida strains, with fractional inhibitory concentration indices ranging from 0.28 to 0.5. Time killing studies revealed a >2-log reduction in the viability of Candida albicans ATCC 10231 cells after 3 h incubation with the combination amphotericin B plus bacillomycin D, at their subinhibitory concentration. Interestingly, when the two drugs were used together at those dosages displaying a synergism in the anti-Candida activity, no cytotoxic effect was observed against mammalian cells. Therefore, the combination bacillomycin D/amphotericin B may represent a valid alternative to conventional antifungals for topical treatment of C. albicans infections. To the best of our knowledge, this is the first report describing the in vitro interaction between the antifungal drug amphotericin B and bacillomycin D against pathogenic Candida species.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
MOPS
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG