  • Search Results
  • Utilization of proteomics in experimental field conditions--A case study of poplars growing on grassland affected by long-term starch wastewater irrigation.
Utilization of proteomics in experimental field conditions--A case study of poplars growing on grassland affected by long-term starch wastewater irrigation.
Journal of proteomics (2015-06-14)
Agnieszka Szuba, Gabriela Lorenc-Plucińska
ABSTRACT
The presented study verified the possibility of using proteomics as a tool for investigating poplars growing on obviously separate plots. The examination covered poplars planted on grassland irrigated for 40 years with potato industry wastewater and in a plot appropriate for poplar planting, spaced at a distance of 67 km from each other (hereinafter referred to as forest). The work aimed to compare the obtained proteomic results with data on biometric and biochemical parameters and mineral composition as well as to assess, at a molecular level, the usefulness of grasslands for planting. Proteome analysis showed that most of the stress-related proteins detected were less abundant on the irrigated grassland, confirming the viability of its revegetation with poplars. Proteomic data corresponded well with the other results, highlighting the probable reason for the proteome changes; i.e. deficiency of phosphate ions detected in the forest area. Moreover, proteome analysis revealed biotic stress symptoms in plants growing on the grassland, which were also well explained by other data but would not have been detected without performing the proteomic analysis. Therefore, environmental plant proteomics is a useful and valuable tool during field studies, even when samples are taken from plots some distance apart.
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
SAFC
Iodoacetamide
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard