MilliporeSigma
Health-Related Quality of Life of Former Lead Workers in Brazil.
International journal of environmental research and public health (2015-11-06)
Martha Carvalho Pereira Teixeira, Fernando Martins Carvalho, Liliane Lins
ABSTRACT
Little is known about the health-related quality of life of former lead workers. Using the Short-Form 36 Questionnaire (SF-36), a cross-section design study evaluated the health-related quality of life of 186 former workers of a lead smelter that operated in Santo Amaro da Purificação, Brazil, from 1960 to 1993, when it closed down. The smelter had very poor occupational and environmental hygiene standards. The health-related quality of life of former lead workers was low, compared to population-based and other nosological groups from Brazil. Former lead workers who indicated metal poisoning, difficulty getting another job and who could not get another job after dismissal by the smelter presented poorer health-related quality of life. Former lead workers with poor health-related quality of life form part of the huge occupational liability left by the Santo Amaro lead smelter.
Sigma-Aldrich
Mercury, electronic grade, 99.9999% trace metals basis
Sigma-Aldrich
Mercury, ≥99.99% trace metals basis
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis