MilliporeSigma
Ozone detoxification of steam-pretreated Norway spruce.
Biotechnology for biofuels (2015-12-01)
Adnan Cavka, Anna Wallenius, Björn Alriksson, Nils-Olof Nilvebrant, Leif J Jönsson
ABSTRACT
Pretreatment of lignocellulose for biochemical conversion commonly results in formation of by-products that inhibit microorganisms and cellulolytic enzymes. To make bioconversion processes more efficient, inhibition problems can be alleviated through conditioning. Ozone is currently commercially employed in pulp and paper production for bleaching, as it offers the desirable capability to disrupt unsaturated bonds in lignin through an ionic reaction known as ozonolysis. Ozonolysis is more selective towards lignin than cellulose, for instance, when compared to other oxidative treatment methods, such as Fenton's reagent. Ozone may thus have desirable properties for conditioning of pretreated lignocellulose without concomitant degradation of cellulose or sugars. Ozone treatment of SO2-impregnated steam-pretreated Norway spruce was explored as a potential approach to decrease inhibition of yeast and cellulolytic enzymes. This novel approach was furthermore compared to some of the most effective methods for conditioning of pretreated lignocellulose, i.e., treatment with alkali and sodium dithionite. Low dosages of ozone decreased the total contents of phenolics to about half of the initial value and improved the fermentability. Increasing ozone dosages led to almost proportional increase in the contents of total acids, including formic acid, which ultimately led to poor fermentability at higher ozone dosages. The decrease of the contents of furfural and 5-hydroxymethylfurfural was inversely proportional (R (2) > 0.99) to the duration of the ozone treatment, but exhibited no connection with the fermentability. Ozone detoxification was compared with other detoxification methods and was superior to treatment with Fenton's reagent, which exhibited no positive effect on fermentability. However, ozone detoxification was less efficient than treatment with alkali or sodium dithionite. High ozone dosages decreased the inhibition of cellulolytic enzymes as the glucose yield was improved with 13 % compared to that of an untreated control. Low dosages of ozone were beneficial for the fermentation of steam-pretreated Norway spruce, while high dosages decreased the inhibition of cellulolytic enzymes by soluble components in the pretreatment liquid. While clearly of interest for conditioning of lignocellulosic hydrolysates, future challenges include finding conditions that provide beneficial effects both with regard to enzymatic saccharification and microbial fermentation.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Levulinic acid, ≥97%, FG
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets