  • Search Results
  • Imatinib mesylate lacks efficacy in relapsed/refractory peripheral T cell lymphoma.
Imatinib mesylate lacks efficacy in relapsed/refractory peripheral T cell lymphoma.
Leukemia & lymphoma (2014-07-12)
Eric Jacobsen, Olga Pozdnyakova, Robert Redd, David C Fisher, David M Dorfman, Paola Dal Cin, Ann LaCasce, Philippe Armand, Ephraim Hochberg, Gregory Cote, Ali Shahsafaei, Donna Neuberg, Jennifer R Brown, Arnold S Freedman
ABSTRACT
Platelet derived growth factor-α (PDGFR-α) is expressed in peripheral T cell lymphoma, not otherwise specified (PTCL, NOS). Imatinib mesylate demonstrated in vitro cytotoxicity against primary PTCL, NOS cells. We initiated a trial of imatinib in 12 patients with relapsed or refractory T-cell non-Hodgkin lymphoma (T-NHL). PDGFR-α expression by immunohistochemistry and fluorescence in situ hybridization (FISH) to assess for FIP1L1-PDGFR-α fusion and/or PDGFR-α amplification were not required for study entry. We documented no objective responses. The median progression-free survival was 21.0 days (90% confidence interval [CI] 15.0, 28.0) and median overall survival was 154 days (90% CI 35, 242). Four patients had tissue available for analysis of PDGFR-α by immunohistochemistry and three of these patients' tumors expressed PDGFR-α. Imatinib was not effective for the treatment of peripheral T cell lymphoma in an unselected group of patients in which PDGFR-α expression was not required for study entry.
Sigma-Aldrich
3,3′-Diaminobenzidine, ≥97% (HPLC)
Sigma-Aldrich
3,3′-Diaminobenzidine, 98%