  • Search Results
  • Effects of light-emitting diode irradiation on RANKL-induced osteoclastogenesis.
Effects of light-emitting diode irradiation on RANKL-induced osteoclastogenesis.
Lasers in surgery and medicine (2015-09-24)
HongMoon Sohn, Youngjong Ko, Mineon Park, Donghwi Kim, Young Lae Moon, Yeon Joo Jeong, Hyeonjun Lee, Yeonhee Moon, Byung-Chul Jeong, Okjoon Kim, Wonbong Lim
ABSTRACT
Bone homeostasis is maintained by a balance between osteoblastic bone formation and osteoclastic bone resorption, where intracellular reactive oxygen species (ROS) are crucial mediators of osteoclastogenesis. Recently, low-level light therapy (LLLT), a form of laser medicine used in various clinical fields, was shown to alleviate oxidative stress by scavenging intracellular ROS. The present study aimed to investigate the impact of 635 nm irradiation from a light-emitting diode (LED) on osteoclastogenesis from receptor activator of nuclear factor kappa-B (NF-κB) ligand (RANKL)-stimulated mouse bone marrow-derived macrophages (BMMs). The effects of LED irradiation on osteoclastogenesis were assessed in tartrate-resistant acid phosphatase (TRAP), 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), cell viability, and resorption pit formation, respectively. Quantitative real-time polymerase chain reaction (qPCR) and Western blot analyses were also performed to assess mRNA expression of osteoclastogenesis-related genes and phosphorylation of extracellular signal-regulated kinase 1/2 (ERK 1/2), p38, and c-Jun-N-terminal kinase (JNK). NF-κB activity was assayed by luciferase reporter assay and Intracellular ROS generation was investigated by the 2',7'-dichlorodihydrofluorescein diacetate (H2 DCF-DA) detection method. LED irradiation significantly inhibited RANKL-mediated osteoclast differentiation from BMMs and mRNA expression of TRAP, osteoclast-associated immunoglobulin-like receptor (OSCAR), and dendrocyte-expressed seven-transmembrane protein (DC-STAMP). Exposure to LED light likewise significantly decreased RANKL-facilitated NF-κB activity, p38 and ERK phosphorylation and intracellular ROS generation, and increased gene expression of nuclear factor E2-related factor 2 (Nrf2). Taken together, the results presented herein show that LED irradiation downregulates osteoclastogenesis by reducing ROS production. Therefore, LED irradiation/LLLT might be useful as an alternative, conservative approach to osteoporosis management.
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Calcium phosphate, purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)