  • Search Results
  • A carbon needle microelectrode decorated with TiO₂ nanosheets dominated by reactive facets as a highly electrocatalytic sensing element.
A carbon needle microelectrode decorated with TiO₂ nanosheets dominated by reactive facets as a highly electrocatalytic sensing element.
Talanta (2015-06-17)
Xue Bai, Xin Huang, Qi Zhang, Zulin Hua, Caidie Qin, Qin Qin
ABSTRACT
We demonstrate a highly efficient electrocatalytic activity of TiO2 nanosheets with reactive (001) facets grown on a carbon needle microelectrode (TiO2-CNE). This microelectrode is prepared by a simple hydrothermal process. Scanning electron microscopy, high-resolution transmission electron microscopy, and electrochemical impedance spectroscopy were used to characterize as-prepared TiO2-CNE. These methods confirm that the surface area-to-volume ratio and electron transfer rate of TiO2-CNE are considerably greater than those of the bare electrode. In addition, the amperometric response for acetaminophen (AP) oxidation is remarkably facilitated by 13.9 times at the TiO2-CNE by the enlarged surface area-to-volume ratio, as well as the exposure of high-energy (001) facets. Furthermore, the constructed electrochemical sensor displays excellent analytical performance for AP detection over a range of 0.002-100.0 μM, with a detection limit of 0.83 nM (S/N=3). The electrode was applied for the in vitro determination of AP in fish blood 30 min after injection in the abdominal cavity. Consequently, the present approach shows feasibility and great potential in constructing highly sensitive and low-consumption miniaturized electrochemical sensors in the field of biological applications.
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.