  • Search Results
  • Dopamine-functionalized InP/ZnS quantum dots as fluorescence probes for the detection of adenosine in microfluidic chip.
Dopamine-functionalized InP/ZnS quantum dots as fluorescence probes for the detection of adenosine in microfluidic chip.
International journal of nanomedicine (2015-09-09)
Seshadri Reddy Ankireddy, Jongsung Kim
ABSTRACT
Microbeads are frequently used as solid supports for biomolecules such as proteins and nucleic acids in heterogeneous microfluidic assays. Chip-based, quantum dot (QD)-bead-biomolecule probes have been used for the detection of various types of DNA. In this study, we developed dopamine (DA)-functionalized InP/ZnS QDs (QDs-DA) as fluorescence probes for the detection of adenosine in microfluidic chips. The photoluminescence (PL) intensity of the QDs-DA is quenched by Zn(2+) because of the strong coordination interactions. In the presence of adenosine, Zn(2+) cations preferentially bind to adenosine, and the PL intensity of the QDs-DA is recovered. A polydimethylsiloxane-based microfluidic chip was fabricated, and adenosine detection was confirmed using QDs-DA probes.
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Supelco
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%