  • Search Results
  • Identification and characterization of a stachyose synthase gene controlling reduced stachyose content in soybean.
Identification and characterization of a stachyose synthase gene controlling reduced stachyose content in soybean.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik (2015-07-17)
Dan Qiu, Tri Vuong, Babu Valliyodan, Haiying Shi, Binhui Guo, J Grover Shannon, Henry T Nguyen
ABSTRACT
We identified and characterized a mutant of soybean stachyose synthase gene controlling reduced stachyose content which benefit the soybean seed composition breeding program in the future. It has been shown that in soybean, increased sucrose and reduced raffinose family oligosaccharides would have a positive impact on the world's feed industry by improving digestibility and feed efficiency. We searched for new sources of modified oligosaccharide content in a subset of the USDA Soybean Germplasm Collection and then identified plant introduction (PI) 603176A as having ultra-low stachyose content (0.5%). We identified a 33-bp deletion mutant in the putative stachyose synthase gene (STS gene, Glyma19g40550) of PI 603176A. A co-dominate indel marker was successfully developed from this 33-bp deletion area and was genetically mapped into two F 2:3 populations and a F 4:5 population, which associated with low stachyose content in the progeny lines. These observations provided strong evidence that the STS gene is responsible for stachyose biosynthesis in the soybean plant. Expression of the sts gene remained at the normal level, suggesting the loss of function in the gene is due to defective protein function. This gene-based perfect genetic marker for low stachyose content can be useful for marker-assisted selection in soybean molecular breeding programs.
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications