  • Search Results
  • Characterization of fish gelatin-gum arabic complex coacervates as influenced by phase separation temperature.
Characterization of fish gelatin-gum arabic complex coacervates as influenced by phase separation temperature.
International journal of biological macromolecules (2015-06-10)
Mohammad Anvari, Cheol-Ho Pan, Won-Byong Yoon, Donghwa Chung
ABSTRACT
The rheological and structural characteristics of fish gelatin (FG)-gum arabic (GA) complex coacervate phase, separated from an aqueous mixture of 1% FG and 1% GA at pH 3.5, were investigated as influenced by phase separation temperature. Decreasing the phase separation temperature from 40 to 10 °C lead to: (1) the formation of a coacervate phase with a larger volume fraction and higher biopolymer concentrations, which is more viscous, more structural resistant at low shear rates, more shear-thinning at high shear rates, and more condensed in microstructure, (2) a solid-like elastic behavior of the phase separated at 10 °C at a high oscillatory frequency, (3) the increase in gelling and melting temperatures of the coacervate phase (3.7-3.9 °C and 6.2-6.9 °C, respectively), (4) the formation of a more rigid and thermo-stable coacervate gel. The coacervate phase is regarded as a homogeneously networked biopolymer matrix dispersed with water vacuoles and its gel as a weak physical gel reinforced by FG-GA attractive electrostatic interactions.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Rhodamine B isothiocyanate, mixed isomers, BioReagent, suitable for protein labeling
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Rhodamine B isothiocyanate, mixed isomers
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis