  • Search Results
  • Effect of polar and movable (OH or NH2 groups) on the photocatalytic H2 production of alkyl-alkanolamine: a comparative study.
Effect of polar and movable (OH or NH2 groups) on the photocatalytic H2 production of alkyl-alkanolamine: a comparative study.
Environmental technology (2015-03-03)
A F Alkaim, R Dillert, D W Bahnemann
ABSTRACT
The photocatalytic production of molecular hydrogen (H2) from aqueous solutions of alkyl-alkanolamines (triethanolamine (tri-EOA), diethanolamine (di-EOA), ethanolamine (EOA), ethylamine (EA) and ethanol over Pt-loaded commercial (Sachtleben Hombikat UV100) nanoparticle titanium dioxide photocatalysts was studied. These photocatalysts were characterized using X-ray diffraction, Brunauer-Emmett-Teller, field-emission scanning electron microscopy, atomic force microscopy and UV-vis diffuse reflectance spectroscopy techniques. Effect of pH and temperature on photocatalytic hydrogen production of alkyl-alkanolamines was investigated. It was found for all molecules under study that the amount of hydrogen produced at a constant illumination time is a function of pH and temperature. At all temperatures investigated the rate of hydrogen production at pH 9 followed the order tri-EOAethanolamine > di-EOA ≈ EOA > ethanol > EA. From the values of the photocatalytic rate constants at various temperatures, the apparent activation energies were determined for the hydrogen production. The observed dependence of the reaction rate on solution temperature cannot be related to light-driven reaction steps, because the band-gap energy of the semiconductor is too high for thermal excitation to become significant in the temperature range investigated.
Sigma-Aldrich
Ethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethylamine solution, 66.0-72.0% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Potassium hydroxide on alumina
SAFC
Triethanolamine
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Ethylamine solution, 2.0 M in THF
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Chloroplatinic acid hexahydrate, BioXtra
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethylamine, 97%
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Ethylamine, 99.0%