  • Search Results
  • In Vivo Correlation of Glucose Metabolism, Cell Density and Microcirculatory Parameters in Patients with Head and Neck Cancer: Initial Results Using Simultaneous PET/MRI.
In Vivo Correlation of Glucose Metabolism, Cell Density and Microcirculatory Parameters in Patients with Head and Neck Cancer: Initial Results Using Simultaneous PET/MRI.
PloS one (2015-08-14)
Matthias Gawlitza, Sandra Purz, Klaus Kubiessa, Andreas Boehm, Henryk Barthel, Regine Kluge, Thomas Kahn, Osama Sabri, Patrick Stumpp
ABSTRACT
To demonstrate the feasibility of simultaneous acquisition of 18F-FDG-PET, diffusion-weighted imaging (DWI) and T1-weighted dynamic contrast-enhanced MRI (T1w-DCE) in an integrated simultaneous PET/MRI in patients with head and neck squamous cell cancer (HNSCC) and to investigate possible correlations between these parameters. 17 patients that had given informed consent (15 male, 2 female) with biopsy-proven HNSCC underwent simultaneous 18F-FDG-PET/MRI including DWI and T1w-DCE. SUVmax, SUVmean, ADCmean, ADCmin and Ktrans, kep and ve were measured for each tumour and correlated using Spearman's ρ. Significant correlations were observed between SUVmean and Ktrans (ρ = 0.43; p ≤ 0.05); SUVmean and kep (ρ = 0.44; p ≤ 0.05); Ktrans and kep (ρ = 0.53; p ≤ 0.05); and between kep and ve (ρ = -0.74; p ≤ 0.01). There was a trend towards statistical significance when correlating SUVmax and ADCmin (ρ = -0.35; p = 0.08); SUVmax and Ktrans (ρ = 0.37; p = 0.07); SUVmax and kep (ρ = 0.39; p = 0.06); and ADCmean and ve (ρ = 0.4; p = 0.06). Simultaneous 18F-FDG-PET/MRI including DWI and T1w-DCE in patients with HNSCC is feasible and allows depiction of complex interactions between glucose metabolism, microcirculatory parameters and cellular density.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems