  • Search Results
  • Inhibition of tumor progression by oral piceatannol in mouse 4T1 mammary cancer is associated with decreased angiogenesis and macrophage infiltration.
Inhibition of tumor progression by oral piceatannol in mouse 4T1 mammary cancer is associated with decreased angiogenesis and macrophage infiltration.
The Journal of nutritional biochemistry (2015-08-25)
Hyerim Song, Jae In Jung, Han Jin Cho, Song Her, Seung-Hae Kwon, Rina Yu, Young-Hee Kang, Ki Won Lee, Jung Han Yoon Park
ABSTRACT
Piceatannol, a polyphenol which exhibits anticancer activities, is found in grapes, red wine and berries. It has been shown to inhibit several transcription factor pathways. The present study was conducted to determine whether oral administration of piceatannol inhibits mammary tumor progression. 4T1 mammary carcinoma cells were injected into the mammary fat pad of syngeneic female BALB/c mice. Starting 1 day later, piceatannol (10- or 20-mg/kg body weight/day) was administered by oral gavage for 30 days. Piceatannol treatment reduced tumor growth. In tumor tissues, piceatannol treatment reduced the expression of transcription factors P-NFκB p65, P-STAT3 and HIF-1α and multiple proteins involved in regulation of cell cycle progression (Ki67, cyclin D1, cyclin A, CDK2, CDK4), angiogenesis (VEGF-A, VEGFR-2, VE-cadherin, CD31) and lymphangiogenesis (VEGF-C, LYVE-1), as well as macrophage infiltration. Piceatannol significantly increased apoptotic cells and expression of both Bax and cleaved caspase-3 but reduced Bcl-2 expression in tumor tissues. In addition, piceatannol reduced the number and volume of pulmonary tumor nodules and expression of MMP-9 in both lung and tumor. It also reduced tissue levels of cytokines/chemokines, including M-CSF and MCP-1. In vitro results revealed that piceatannol inhibited migration of 4T1 cells and monocytes, as well as secretion of MCP-1 and M-CSF by 4T1 cells. 4T1 cell-conditioned medium stimulated monocyte migration, which was suppressed by a CCR2 antibody. These results indicate that alteration in tumor microenvironment (macrophages, transcription factors, etc.) is an important mechanism by which piceatannol inhibits tumor proliferation, angiogenesis and lymphangiogenesis, leading to suppression of mammary tumor growth and metastasis.
Sigma-Aldrich
2,3-Diaminonaphthalene, ≥95% (HPLC), powder
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
2,3-Diaminonaphthalene, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
Nitric oxide, 98.5%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology