  • Search Results
  • Detection of Helicobacter pylori DNA in inflamed dental pulp specimens from Japanese children and adolescents.
Detection of Helicobacter pylori DNA in inflamed dental pulp specimens from Japanese children and adolescents.
Journal of medical microbiology (2014-10-22)
Yuko Ogaya, Ryota Nomura, Yoshiyuki Watanabe, Kazuhiko Nakano
ABSTRACT
The oral cavity has been implicated as a source of Helicobacter pylori infection in childhood. Various PCR methods have been used to detect H. pylori DNA in oral specimens with various detection rates reported. Such disparity in detection rates complicates the estimation of the true infection rate of H. pylori in the oral cavity. In the present study, we constructed a novel PCR system for H. pylori detection and used it to analyse oral specimens. Firstly, the nucleotide alignments of genes commonly used for H. pylori detection were compared using the complete genome information for 48 strains registered in the GenBank database. Candidate primer sets with an estimated amplification size of approximately 300-400 bp were selected, and the specificity and sensitivity of the detection system using each primer set were evaluated. Five sets of primers targeting ureA were considered appropriate, of which a single primer set was chosen for inclusion in the PCR system. The sensitivity of the system was considered appropriate and its detection limit established as one to ten cells per reaction. The novel PCR system was used to examine H. pylori distribution in oral specimens (40 inflamed pulp tissues, 40 saliva samples) collected from Japanese children, adolescents and young adults. PCR analysis revealed that the detection rate of H. pylori in inflamed pulp was 15 %, whereas no positive reaction was found in any of the saliva specimens. Taken together, our novel PCR system was found to be reliable for detecting H. pylori. The results obtained showed that H. pylori was detected in inflamed pulp but not saliva specimens, indicating that an infected root canal may be a reservoir for H. pylori.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles