MilliporeSigma
ANO1 (TMEM16A) in pancreatic ductal adenocarcinoma (PDAC).
Pflugers Archiv : European journal of physiology (2014-08-29)
D R P Sauter, I Novak, S F Pedersen, E H Larsen, E K Hoffmann
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) has one of the worst survival rates of all cancers. ANO1 (TMEM16A) is a recently identified Ca(2+)-activated Cl(-) channel (CaCC) that is upregulated in several tumors. Although ANO1 was subject to extensive studies in the recent years, its pathophysiological function has only been poorly understood. The aim of the present study is to establish the significance of ANO1 in PDAC behavior and demarcate its roles in PDAC from those of the volume-regulated anion channel (VRAC). We performed qPCR and Western blot measurements on different PDAC cell lines (Panc-1, Mia PaCa 2, Capan-1, AsPC-1, BxPC-3) and compared the results to those obtained in a human pancreatic ductal epithelium (HPDE) cell line. All cancer cell lines showed an upregulation of ANO1 on mRNA and protein levels. Whole-cell patch-clamp recordings identified large Ca(2+) and voltage-dependent Cl(-) currents in PDAC cells. Using siRNA knockdown of ANO1 and three ANO1 inhibitors (T16Ainh-A01, CaCCinh-A01, and NS3728), we found that ANO1 is the main constituent of CaCC current in PDAC cells. We further characterized these three inhibitors and found that they had unspecific effects on the free intracellular calcium concentration. Functional studies on PDAC behavior showed that surprisingly inhibition of ANO1 did not influence cellular proliferation. On the other hand, we found ANO1 channel to be pivotal in PDAC cell migration as assessed in wound healing experiments.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetamide, sublimed, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)