  • Search Results
  • Aminoglycoside analysis in food of animal origin with a zwitterionic stationary phase and liquid chromatography-tandem mass spectrometry.
Aminoglycoside analysis in food of animal origin with a zwitterionic stationary phase and liquid chromatography-tandem mass spectrometry.
Analytica chimica acta (2015-06-05)
Cristina Díez, Davy Guillarme, Aline Staub Spörri, Emmanuelle Cognard, Didier Ortelli, Patrick Edder, Serge Rudaz
ABSTRACT
In this study, fourteen highly polar aminoglycoside (AGs) antibiotics were selected. Various stationary phases were tested, including Obelisc R, ZIC-HILIC, BEH amide and aminopropyl. The nature of the stationary phase, mobile phase (water or buffer solutions and acetonitrile), pH (percentage of formic acid), gradient conditions and injection solvents were systematically studied as relevant parameters for tuning retention selectivity and detectability of AGs in liquid chromatography electrospray tandem mass spectrometry (LC-(ESI)-MS/MS). Only the two zwitterionic phases (Obelisc R and ZIC-HILIC) achieved a proper chromatographic separation considering interferences due to the crosstalk effect in low resolution mass spectrometers. The water/acetonitrile mobile phase containing 1% formic acid used with Obelisc R provided more sensitivity than the highly concentrated buffered mobile phases required for ZIC-HILIC. A solid phase extraction (SPE) clean-up procedure with polymeric weak cation exchange (WCX) cartridges was optimized for honey, milk and liver samples. Different brands of cartridges and elution solvents were tested, and the Taurus WCX offered the best recovery rate with a buffer elution at pH 3. The final optimized method was validated in these matrices according to Decision 2002/657/EC. A monitoring campaign for sixty honey, milk and liver samples was carried out at the Food Authority Control in Geneva. The concentration of dihydrostreptomycin (DSTP) found in one ovine liver exceeded the established maximum residue levels (MRLs) within the European and Swiss legislations but it was compliant taking into account the validation data.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, potency: ≥603 μg per mg
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, powder, γ-irradiated
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Amikacin hydrate, aminoglycoside antibiotic
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Paromomycin sulfate salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%