  • Search Results
  • Novel antiapoptotic effect of TBX15: overexpression of TBX15 reduces apoptosis in cancer cells.
Novel antiapoptotic effect of TBX15: overexpression of TBX15 reduces apoptosis in cancer cells.
Apoptosis : an international journal on programmed cell death (2015-07-29)
Jéssica Arribas, Esteban Giménez, Ricard Marcos, Antonia Velázquez
ABSTRACT
T-box genes regulate development processes, some of these genes having also a role in cell proliferation and survival. TBX15 is a T-box transcription factor that, recently, has been proposed as a marker in prostate cancer, but its function in carcinogenesis is unknown. Here the role of TBX15 in carcinogenesis was investigated using thyroid cancer cell lines. First, using western blot analysis, we show that the expression of TBX15 was altered in thyroid cancer cells lines with respect to normal thyroid cells. Transfection of thyroid cancer cells with TBX15, in the presence or absence of camptothecin as a cytotoxic agent, proved non effect of TBX15 in cell viability; but, it increased cell proliferation after 48 h of transfection (P < 0.01). Consistently, apoptosis was reduced in TBX15 transfected cells (P < 0.01) which also showed a decrease of the proapoptotic Bax regulator and an increase of the antiapoptotic Bcl2 and Bcl-XL regulators. Additionally, siRNA shutdown of constitutive TBX15 increased apoptosis. TBX15 transfection did not alter colony formation and cell migration. Taken together, these results indicate for the first time an antiapoptotic role of TBX15 in cancer cells, suggesting a contribution of TBX15 in carcinogenesis and the potential therapeutic target of TBX15.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%