  • Search Results
  • Synthesis of iron oxyhydroxide-coated rice straw (IOC-RS) and its application in arsenic(V) removal from water.
Synthesis of iron oxyhydroxide-coated rice straw (IOC-RS) and its application in arsenic(V) removal from water.
Journal of water and health (2015-09-01)
Igor W K Ouédraogo, Erol Pehlivan, Hien T Tran, Yvonne L Bonzi-Coulibaly, Dieter Zachmann, Müfit Bahadir
ABSTRACT
Because of the recognition that arsenic (As) at low concentrations in drinking water causes severe health effects, the technologies of As removal have become increasingly important. In this study, a simplified and effective method was used to immobilize iron oxyhydroxide onto a pretreated naturally occurring rice straw (RS). The modified RS adsorbent was characterized, using scanning electron microscope, Fourier transform infrared spectroscopy, thermogravimetric analyzer, and surface area analyzer. Experimental batch data of As(V) adsorption were modeled by the isotherms and kinetics models. Although all isotherms, the Langmuir model fitted the equilibrium data better than Freundlich and Dubinin-Radushkevich models and confirmed the surface homogeneity of adsorbent. The iron oxyhydroxide-coated rice straw (IOC-RS) was found to be effective for the removal of As(V) with 98.5% sorption efficiency at a concentration of <50 mg/L of As(V) solution, and thus maximum uptake capacity is ∼22 and 20 mg As(V)/g of IOC-RS at pH 4 and 6, respectively. The present study might provide new avenues to achieve the As concentrations required for drinking water recommended by the World Health Organization.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sulfuric acid, 99.999%