  • Search Results
  • Ultrastructural evaluation of mesenchymal stem cells from inflamed periodontium in different in vitro conditions.
Ultrastructural evaluation of mesenchymal stem cells from inflamed periodontium in different in vitro conditions.
Microscopy research and technique (2015-07-17)
Raluca Zaganescu, Lucian Barbu Tudoran, Emoke Pall, Adrian Florea, Alexandra Roman, Andrada Soanca, Carmen Mihaela Mihu
ABSTRACT
This research aimed to observe the behavior of mesenchymal stem cells (MSCs) isolated from periodontal granulation tissue (gt) when manipulated ex vivo to induce three-dimensional (3D) spheroid (aggregates) formation as well as when seeded on two bone scaffolds of animal origin. Periodontal gt was chosen as a MSC source because of its availability, considering that it is eliminated as a waste material during conventional surgical therapies. 3D aggregates of cells were generated; they were grown for 3 and 7 days, respectively, and then prepared for transmission electron microscopic analysis. The two biomaterials were seeded for 72 h with gtMSCs and prepared for scanning electronic microscopic observation. The ultrastructural analysis of 3D spheroids remarked some differences between the inner and the outer cell layers, with a certain commitment observed at the inner cells. Both scaffolds showed a relatively smooth surface at low magnification. Macro- and micropores having a scarce distribution were observed on both bone substitutes. gtMSCs grew with relative difficulty on the biomaterials. After 72 h of proliferation, gtMSCs scarcely covered the surface of bovine bone scaffolds, demonstrating fibroblast-like or star-like shapes with elongated filiform extensions. Our results add other data on the possible usefulness of gtMSC and could question the current paradigm regarding the complete removal of chronically inflamed gts from the defects during periodontal surgeries. Until optimal protocols for ex vivo manipulation of MSCs are available for clinical settings, it is advisable to use biocompatible bone substitutes that allow the development of progenitor cells.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O