MilliporeSigma
Improved protease digestion conditions for membrane protein detection.
Electrophoresis (2015-04-18)
Lie Min, Leila H Choe, Kelvin H Lee
ABSTRACT
This work presents improved protease digestion conditions for membrane protein detection. The enzymatic digest of bacteriorhodopsin (BR), a model membrane protein with seven transmembrane domains (TMDs) was investigated. An initial in-gel digestion identified 17% BR sequence coverage, including part of the seventh TMD. To improve sequence coverage, BR digest was tested with different concentrations of RapiGest, methanol (MeOH) and SDS using either trypsin or chymotrypsin. Two improved conditions, 0.01% SDS or the combination of 10% MeOH and 0.01% RapiGest, were chosen. Trypsin digestions in both conditions achieved more than 40% BR sequence coverage compared to 0% using standard digestion conditions. Peptides detected from trypsin and chymotrypsin digestions in the same condition were combined to maximize sequence coverage. The same conditions were applied to a different membrane protein with one TMD, Selenoprotein S, and proteins from Escherichia coli. For Selenoprotein S, a higher sequence coverage, including a peptide from the TMD, was detected from the improved condition compared to the typical condition. The application of both improved conditions to a membrane protein fraction of Escherichia coli resulted in the identification of 309 (SDS) and 329 (MeOH/RapiGest) unique proteins of which 140/309 and 148/329 were membrane proteins.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis