  • Search Results
  • Quantitative Assessment of Destruxins from Strawberry and Maize in the Lower Parts per Billion Range: Combination of a QuEChERS-Based Extraction Protocol with a Fast and Selective UHPLC-QTOF-MS Assay.
Quantitative Assessment of Destruxins from Strawberry and Maize in the Lower Parts per Billion Range: Combination of a QuEChERS-Based Extraction Protocol with a Fast and Selective UHPLC-QTOF-MS Assay.
Journal of agricultural and food chemistry (2015-05-26)
Judith Taibon, Sonja Sturm, Christoph Seger, Hermann Strasser, Hermann Stuppner
ABSTRACT
The entomopathogenic fungus Metarhizium brunneum is widely applied as a biological pest control agent. Consequently, its use has to be accompanied by a risk management approach, which includes the need to monitor the fate of its bioactive metabolites in the environment, for example, in treated crops. A fast and selective UHPLC-QTOF-MS method was developed to monitor the presence of secreted destruxins in two model food plants for the application of this fungal biocontrol agent, namely, strawberry and maize. The liquid chromatography-mass spectrometric assay for destruxin trace analysis is combined with a novel QuEChERS-based extraction protocol. The whole assay was optimized for the application in these crops, and it allows quantitative analysis of the major M. brunneum metabolites destruxin A, 1, destruxin B, 2, and destruxin E, 3, down to the parts per billion range. In strawberry, limits of quantitation (LOQs) were found to be <2.0 ppb for all analytes; in maize LOQs were found to be <3.2 ppb for destruxin A and destruxin B. Destruxin E showed a distinctive loss of recovery in maize and was excluded from further quantitative analysis in this crop. For both crops assay linearities ranged from the LOQs to 100 ppb, interassay repeatabilities (RSD) were found to be better than 16.4%, and accuracies ranged from 83.5 to 105.3% (assessed at four spiking levels between 5 and 75 ppb).
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Dichloromethane, ≥99.9%
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Roxithromycin, 95.0-102.0% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis