  • Search Results
  • Incorporation of porcine adenovirus 4 fiber protein enhances infectivity of adenovirus vector on dendritic cells: implications for immune-mediated cancer therapy.
Incorporation of porcine adenovirus 4 fiber protein enhances infectivity of adenovirus vector on dendritic cells: implications for immune-mediated cancer therapy.
PloS one (2015-05-02)
Ivy Wilkinson-Ryan, Julius Kim, Sojung Kim, Ferhat Ak, Lindzy Dodson, Marco Colonna, Matthew Powell, David Mutch, Dirk Spitzer, Ted Hansen, Simon P Goedegebuure, David Curiel, William Hawkins
ABSTRACT
One strategy in cancer immunotherapy is to capitalize on the key immunoregulatory and antigen presenting capabilities of dendritic cells (DCs). This approach is dependent on efficient delivery of tumor specific antigens to DCs, which subsequently induce an anti-tumor T-cell mediated immune response. Human adenovirus serotype 5 (HAdV5) has been used in human studies for gene delivery, but has limited infection in DCs, which lack the proper receptors. Addition of the porcine fiber knob (PK) from porcine adenovirus type 4 to HAdV5 allows the virus to deliver genetic material via binding to glycosylated surface proteins and bypasses the coxsackie-and-adenovirus receptor required by wild-type HAdV5. In this study we explored the potential therapeutic applications of an adenovirus with PK-based tropism against cancers expressing mesothelin. Infectivity and gene transfer assays were used to compare Ad5-PK to wild-type HAdV5. Mouse models were used to demonstrate peptide specificity and T-cell responses. We show that the PK modification highly augmented infection of DCs, including the CD141+ DC subset, a key subset for activation of naïve CD8+ T-cells. We also show that Ad5-PK increases DC infectivity and tumor specific antigen expression. Finally, vaccination of mice with the Ad5-PK vector resulted in enhanced T-cell-mediated interferon gamma (IFN-γ) release in response to both mesothelin peptide and a tumor line expressing mesothelin. Ad5-PK is a promising tool for cancer immunotherapy as it improves infectivity, gene transfer, protein expression, and subsequent T-cell activation in DCs compared to wild-type HAdV5 viruses.
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O