MilliporeSigma
Association between Dietary Magnesium Intake and Hyperuricemia.
PloS one (2015-11-05)
Yi-Lun Wang, Chao Zeng, Jie Wei, Tuo Yang, Hui Li, Zhen-Han Deng, Ye Yang, Yi Zhang, Xiang Ding, Dong-Xing Xie, Tu-Bao Yang, Guang-Hua Lei
ABSTRACT
To examine the cross-sectional associations between dietary magnesium (Mg) intake and hyperuricemia (HU). 5168 subjects were included in this study. Dietary intake was assessed using a validated semi-quantitative food frequency questionnaire. Hyperuricemia (HU) was defined as uric acid ≥ 416 μmol/L for male population and ≥ 360 μmol/L for female. A multivariable logistic analysis model was applied to test the associations after adjusting a number of potential confounding factors. The relative odds of the overall prevalence of HU were decreased by 0.57 times in the fourth quintile of Mg intake (OR 0.57, 95% CI 0.35-0.94) and 0.55 times in the fifth quintile (OR 0.55, 95% CI 0.30-1.01) comparing with the lowest quintile, and P for trend was 0.091. The results of multivariable linear regression also suggested a significant inverse association between serum uric acid and Mg intake (β = -0.028, P = 0.022). For male, the relative odds of HU were decreased by 0.62 times in the third quintile of Mg intake (OR 0.62, 95% CI 0.40-0.97), 0.40 times in the fourth quintile (OR 0.40, 95% CI 0.23-0.72) and 0.35 times in the fifth quintile (OR 0.35, 95% CI 0.17-0.71) comparing with the lowest quintile, and P for trend was 0.006. Multivariable adjusted inverse association was also existed between serum uric acid and Mg intake in male population (β = -0.061, P = 0.002). However, no significant association was observed between dietary Mg intake and HU for female. The findings of this cross-sectional study indicated that dietary Mg intake is inversely associated with HU, independent of some major confounding factors. In addition, this association remains valid for the male subgroup, but not for the female subgroup. LevelIII, cross-sectional study.
Sigma-Aldrich
Uric acid, ≥99%, crystalline
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications