  • Search Results
  • A new approach in the synthesis of La(1-x)Gd(x)FeO3 perovskite nanoparticles--structural and magnetic characterization.
A new approach in the synthesis of La(1-x)Gd(x)FeO3 perovskite nanoparticles--structural and magnetic characterization.
Dalton transactions (Cambridge, England : 2003) (2015-11-04)
R J Wiglusz, K Kordek, M Małecka, A Ciupa, M Ptak, R Pazik, P Pohl, D Kaczorowski
ABSTRACT
A series of highly crystalline orthoferrite nanoparticles (type La(1-x)Gd(x)FeO3, where x = 0 to 1) were prepared using the self-combustion method. Extensive studies including X-ray diffraction, Rietveld refinement and Fourier transform infrared spectroscopy as well as Raman spectroscopy confirmed the orthorhombic space group Pnma of the obtained materials. The calculated average grain size for powders is in the range of 30 to 80 nm. Magnetic characterization of the La(1-x)Gd(x)FeO3 series, performed at 1.72 K, indicated an antiferromagnetic state characterized by some canting of iron magnetic moments, in good agreement with the data reported for similar fine-particle systems.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%