  • Search Results
  • NPM1 activates metabolic changes by inhibiting FBP1 while promoting the tumorigenicity of pancreatic cancer cells.
NPM1 activates metabolic changes by inhibiting FBP1 while promoting the tumorigenicity of pancreatic cancer cells.
Oncotarget (2015-06-13)
Yi Zhu, Minmin Shi, Hao Chen, Jiangning Gu, Jiaqiang Zhang, Baiyong Shen, Xiaxing Deng, Junjie Xie, Xi Zhan, Chenghong Peng
ABSTRACT
The nucleophosmin (NPM1) activates cancer development and progression in many malignant tumors. However, the regulatory role and underlying mechanisms of NPM1 in pancreatic cancer are unknown. In this study, we showed that NPM1 was up-regulated in PDAC, which indicated a poor prognosis. We also identified NPM1 could stimulate aerobic glycolysis and repress fructose-1, 6-bisphosphatase 1 (FBP1) in pancreatic cancer cells. Restoring FBP1 expression partially reversed the tumor-promoting effects of NPM1, while the loss of FBP1 in PDAC tissues was indicative of a poorer prognosis. In sum, NPM1 promotes aerobic glycolysis and tumor progression in patients with pancreatic cancer by inhibiting FBP1.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
SAFC
L-Glutamine
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.