MilliporeSigma
Stability and removal of spironolactone from wastewater.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2015-07-21)
Saleh Sulaiman, Mustafa Khamis, Shlomo Nir, Filomena Lelario, Laura Scrano, Sabino A Bufo, Rafik Karaman
ABSTRACT
Stability and removal of spironolactone (SP) from wastewater produced at Al-Quds University Campus were investigated. Kinetic studies on both pure water and wastewater coming from secondary treatment (activated sludge) demonstrated that the potassium-sparing diuretic (water pill), spironolactone, underwent degradation to its hydrolytic derivative, canrenone, in both media. The first-order hydrolysis rate of SP in activated sludge at 25°C (3.80 × 10(-5) s(-1)) was about 49-fold larger than in pure water (7.4 × 10(-7) s(-1)). The overall performance of the wastewater treatment plant (WWTP) installed in the University Campus was assessed showing that more than 90% of spiked SP was removed together with its newly identified metabolites. In order to look for a technology to supplement or replace ultra-filtration membranes, the effectiveness of adsorption and filtration by micelle-clay filters for removing SP was tested in comparison with activated charcoal. Batch adsorption in aqueous suspensions was well described by Langmuir isotherms, showing a better removal by the micelle-clay material. Filtration of SP water solutions by columns filled with a mixture of sand and a micelle-clay complex showed complete removal of the drug at concentrations higher than in sand/activated-charcoal filled filters.
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, NMR reference standard