  • Search Results
  • Combination of microsatellite instability and BRAF mutation status for subtyping colorectal cancer.
Combination of microsatellite instability and BRAF mutation status for subtyping colorectal cancer.
British journal of cancer (2015-05-15)
T T Seppälä, J P Böhm, M Friman, L Lahtinen, V M J Väyrynen, T K E Liipo, A P Ristimäki, M V J Kairaluoma, I H Kellokumpu, T H I Kuopio, J-P Mecklin
ABSTRACT
The objective of the study was to examine the role of microsatellite instability (MSI) and BRAF(V600E)mutation in colorectal cancer (CRC) by categorising patients into more detailed subtypes based on tumour characteristics. Tumour samples from 762 population-based patients with sporadic CRC were analysed for MSI and BRAF(V600E) by immunohistochemistry. Patient survival was followed-up for a median of 5.2 years. Compared with microsatellite stable (MSS) CRC, MSI was prognostic for better disease-free survival (DFS; 5 years: 85.8% vs 75.3%, 10 years: 85.8% vs 72.9%, P=0.027; HR 0.49, CI 0.30-0.80, P=0.005) and disease-specific survival (DSS; 5 years: 83.2% vs 70.5%; 10 years: 83.2 vs 65.0%, P=0.004). Compared with BRAF wild type, BRAF(V600E) was a risk for poor survival (overall survival; 5 years: 62.3% vs 51.6%, P=0.014; HR 1.43, CI 1.07-1.90, P=0.009), especially in rectal cancer (for DSS, HR: 10.60, CI: 3.04-36.92, P<0.001). The MSS/BRAF(V600E) subtype was a risk for poor DSS (HR: 1.88, CI: 1.06-3.31, P=0.030), but MSI/BRAF(V600E) was a prognostic factor for DFS (HR: 0.42, CI: 0.18-0.96, P=0.039). Among stage I-II patients, the MSS/BRAF(V600E) subtype was independently associated with poor DSS (HR: 5.32, CI: 1.74-16.31, P=0.003). Microsatellite instable tumours were associated with better prognosis compared with MSS. BRAF(V600E) was associated with poor prognosis unless it occurred together with MSI. The MSI/BRAF(V600E) subtype was a favourable prognostic factor compared with the MSS/BRAF wild-type subtype. BRAF(V600E) rectal tumours showed particularly poor prognosis. The MSS/BRAF(V600E) subtype was associated with increased disease-specific mortality even in stage I-II CRC.
Product Description
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene