  • Search Results
  • Mesoporous Silica Nanoparticles Decorated with Carbosilane Dendrons as New Non-viral Oligonucleotide Delivery Carriers.
Mesoporous Silica Nanoparticles Decorated with Carbosilane Dendrons as New Non-viral Oligonucleotide Delivery Carriers.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-09-12)
Ángel Martínez, Elena Fuentes-Paniagua, Alejandro Baeza, Javier Sánchez-Nieves, Mónica Cicuéndez, Rafael Gómez, F Javier de la Mata, Blanca González, María Vallet-Regí
ABSTRACT
A novel nanosystem based on mesoporous silica nanoparticles covered with carbosilane dendrons grafted on the external surface of the nanoparticles is reported. This system is able to transport single-stranded oligonucleotide into cells, avoiding an electrostatic repulsion between the cell membrane and the negatively charged nucleic acids thanks to the cationic charge provided by the dendron coating under physiological conditions. Moreover, the presence of the highly ordered pore network inside the silica matrix would make possible to allocate other therapeutic agents within the mesopores with the aim of achieving a double delivery. First, carbosilane dendrons of second and third generation possessing ammonium or tertiary amine groups as peripheral functional groups were prepared. Hence, different strategies were tested in order to obtain their suitable grafting on the outer surface of the nanoparticles. As nucleic acid model, a single-stranded DNA oligonucleotide tagged with a fluorescent Cy3 moiety was used to evaluate the DNA adsorption capacity. The hybrid material functionalised with the third generation of a neutral dendron showed excellent DNA binding properties. Finally, the cytotoxicity as well as the capability to deliver DNA into cells, was tested in vitro by using a human osteoblast-like cell line, achieving good levels of internalisation of the vector DNA/carbosilane dendron-functionalised material without affecting the cellular viability.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, BioReagent, suitable for fluorescence, ≥96.0% (HPCE)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Supelco
Iodomethane solution, certified reference material, 2000 μg/mL in methanol: water (4:1)
Sigma-Aldrich
3-(Triethoxysilyl)propyl isocyanate, 95%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
Hexadecyltrimethylammonium bromide
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)