  • Search Results
  • Synthesis and characterization of Bodipy functionalized magnetic iron oxide nanoparticles for potential bioimaging applications.
Synthesis and characterization of Bodipy functionalized magnetic iron oxide nanoparticles for potential bioimaging applications.
Colloids and surfaces. B, Biointerfaces (2015-02-25)
Seda Demirel Topel, Önder Topel, R Beklem Bostancıoğlu, A Tansu Koparal
ABSTRACT
Multifunctional magnetic nanoparticles were synthesized for potential bio-imaging applications. Uniform PEI coated magnetic Fe3O4 (PEI-Fe3O4) nanoparticles were prepared by a modified co-precipitation method and then covalently conjugated with a fluorophore molecule, Bodipy-5 by the DCC/DMAP coupling reaction. The covalent binding of Bodipy-5 to the PEI coated magnetic Fe3O4 nanoparticles were confirmed by means of FTIR and XPS measurements. The imaging ability of the Bodipy coated magnetic nanoparticles was determined on two human cancer cells, A549 (human lung adenocarcinoma epithelial) and Ishikawa (endometrial adenocarcinoma), for the first time. Cytotoxicity of BOD-MNPs was evaluated in both cancer cells and healthy human umbilical vein endothelial cell line (HUVEC) by standard MTT (3-(4,5-dimethythiazol-2-yl)-2,5-diphenyl tetrazolium bromide) assay. In vitro activities of the nanoparticles were also investigated.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O