  • Search Results
  • Oxidative Stress Markers and Histological Analysis in Diverse Organs from Rats Treated with a Hepatotoxic Dose of Cr(VI): Effect of Curcumin.
Oxidative Stress Markers and Histological Analysis in Diverse Organs from Rats Treated with a Hepatotoxic Dose of Cr(VI): Effect of Curcumin.
Biological trace element research (2015-03-17)
Wylly Ramsés García-Niño, Zyanya Lucía Zatarain-Barrón, Rogelio Hernández-Pando, Claudia Cecilia Vega-García, Edilia Tapia, José Pedraza-Chaverri
ABSTRACT
Hexavalent chromium [Cr(VI)] compounds are extremely toxic and carcinogenic. Despite the vast quantity of reports about Cr(VI) toxicity, the information regarding its effects when it is intraperitoneally (i.p.) administered is still limited. In contrast, it has been shown that curcumin prevents hepatotoxicity induced by a single intraperitoneal injection of 15 mg/kg body weight (b.w.) of potassium dichromate (K2Cr2O7). This study aims to evaluate oxidative stress markers, the activity of antioxidant enzymes, and the potential histological injury in brain, heart, lung, kidney, spleen, pancreas, stomach, and intestine from rats treated with a hepatotoxic dose of K2Cr2O7 (15 mg/kg b.w.), and the effect of curcumin pretreatment. Rats were divided into four groups: control, curcumin, K2Cr2O7, and curcumin+K2Cr2O7. At the end of the treatment, plasma and ascites fluid were collected and target organs were dissected out for biochemical and histological analysis. K2Cr2O7 induced hepatotoxicity but failed to induce in all the other studied organs either oxidative or histological injury, since levels of malondialdehyde (MDA), glutathione (GSH), and the activity of superoxide dismutase (SOD), catalase (CAT), and related GSH enzymes were unchanged. As expected, curcumin was safe. Lack of K2Cr2O7-induced toxicity in those target organs could be due to the following: (1) route of administration, (2) absorption through the portal circulation, (3) lower dose than needed, (4) short time of exposure, or (5) repeated doses are required to produce damage. Thus, the intraperitoneal injection of 15 mg/kg of K2Cr2O7, that is able to induce hepatotoxicity, was unable to induce histological and oxidative damage in other target organs.
Sigma-Aldrich
Bromocresol Green, ACS reagent, Dye content 95 %
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
MOPS
SAFC
HEPES
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder