  • Search Results
  • Utilizing Ultrasound to Transiently Increase Blood-Brain Barrier Permeability, Modulate of the Tight Junction Proteins, and Alter Cytoskeletal Structure.
Utilizing Ultrasound to Transiently Increase Blood-Brain Barrier Permeability, Modulate of the Tight Junction Proteins, and Alter Cytoskeletal Structure.
Current neurovascular research (2015-08-05)
Mi Jung Bae, Young Mi Lee, Yeoun Hee Kim, Hyung Soo Han, Hak Jong Lee
ABSTRACT
The central nervous system is protected by the blood-brain barrier (BBB). The tight junction (TJ) proteins claudin-5 and zonula occludens-1 (ZO-1) as well as the cytoskeletal component F-actin play key roles in maintaining homeostasis of the BBB. Increases in BBB permeability may be beneficial for the delivery of pharmacological substances into the brain. Therefore, here, we assessed the use of ultrasound to induce transient enhancement of BBB permeability. We used fluorescein isothiocyanate (FITC)-dextran 40 to detect changes in the membrane permeability of bEnd.3 cells during ultrasound treatment. Ultrasound increased FITC-dextran 40 uptake into bEnd.3 cells for 2-6 h after treatment; however, normal levels returned after 24 h. An insignificant increase in lactate dehydrogenase (LDH) leakage also occurred 3 and 6 h after ultrasound treatment, whereas at 24 h, LDH leakage was indistinguishable between the control and treatment groups. Expression of claudin-5, ZO-1, and F-actin at the messenger RNA (mRNA) and protein levels was assessed with real-time polymerase chain reaction and western blotting. Ultrasound induced a transient decrease in claudin-5 mRNA and protein expression within 2 h of treatment; however, no significant changes in ZO-1 and F-actin expression were observed. Claudin-5, ZO-1, and F-actin immunofluorescence demonstrated that the cellular structures incorporating these proteins were transiently impaired by ultrasound. In conclusion, our ultrasound technique can temporarily increase BBB permeability without cytotoxicity to exposed cells, and the method can be exploited in the delivery of drugs to the brain with minimal damage.
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Glycerin, meets USP testing specifications