  • Search Results
  • Maternal immune activation in late gestation increases neuroinflammation and aggravates experimental autoimmune encephalomyelitis in the offspring.
Maternal immune activation in late gestation increases neuroinflammation and aggravates experimental autoimmune encephalomyelitis in the offspring.
Brain, behavior, and immunity (2014-08-12)
Adriano Zager, Jean Pierre Peron, Gregory Mennecier, Sandra C Rodrigues, Thiago P Aloia, João Palermo-Neto
ABSTRACT
Multiple sclerosis (MS) is characterized by an autoimmune response against myelin antigens driven by autoreactive T cells. Several lines of evidence indicate that environmental factors, such as previous infection, can influence and trigger autoimmune responses. However, the importance of the gestational period, particularly under inflammatory conditions, on the modulation of MS and related neuroinflammation by the offspring is unknown. This study aimed to evaluate the impact of prenatal exposure to lipopolysaccharide (LPS) during late gestation on the neuroinflammatory response in primary mixed glial cultures and on the progression of experimental autoimmune encephalomyelitis (EAE, an animal model of MS) in the offspring. LPS (Escherichia coli 0127:B8, 120μg/kg) was administered intraperitoneally to pregnant C57BL/6J mice on gestational day 17, and the offspring were assigned to two experiments: (1) mixed glial cultures generated using the brain of neonates, stimulated in vitro with LPS, and (2) adult offspring immunized with MOG35-55. The EAE clinical symptoms were followed for 30days. Different sets of animals were sacrificed either during the onset (7days post-immunization [p.i.]), when spleen and lymph nodes were collected, or the peak of disease (20days p.i.), when CNS were collected for flow cytometry, cytokine production, and protein/mRNA-expression analysis. The primary CNS cultures from the LPS-treated group produced exaggerated amounts of IL-6, IL-1β and nitrites after in vitro stimulus, while IL-10 production was lowered compared to the data of the control group. Prenatal exposure to LPS worsened EAE disease severity in adult offspring, and this worsening was linked to increased CNS-infiltrating macrophages, Th1 cells and Th17 cells at the peak of EAE severity; additionally, exacerbated gliosis was evidenced in microglia (MHC II) and astrocytes (GFAP protein level and immunoreactivity). The IL-2, IL-6 and IL-17 levels in the spleen and lymph nodes were increased in the offspring of the LPS-exposed dams. Our results indicate that maternal immune activation during late gestation predispose the offspring to increased neuroinflammation and potentiate the autoimmune response and clinical manifestation of EAE.
Sigma-Aldrich
4-Methylumbelliferyl β-D-galactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis