  • Search Results
  • Evidence for synergistic activity of plant-derived essential oils against fungal pathogens of food.
Evidence for synergistic activity of plant-derived essential oils against fungal pathogens of food.
Food microbiology (2015-12-19)
Farah Hossain, Peter Follett, Khang Dang Vu, Mehdi Harich, Stephane Salmieri, Monique Lacroix
ABSTRACT
The antifungal activities of eight essential oils (EOs) namely basil, cinnamon, eucalyptus, mandarin, oregano, peppermint, tea tree and thyme were evaluated for their ability to inhibit growth of Aspergillus niger, Aspergillus flavus, Aspergillus parasiticus and Penicillium chrysogenum. The antifungal activity of the EOs was assessed by the minimum inhibitory concentration (MIC) using 96-well microplate analysis. The interactions between different EO combinations were done by the checkerboard technique. The highest antifungal activity was exhibited by oregano and thyme which showed lower MIC values amongst all the tested fungi. The antifungal activity of the other EOs could be appropriately ranked in a descending sequence of cinnamon, peppermint, tea tree and basil. Eucalyptus and mandarin showed the least efficiency as they could not inhibit any of the fungal growth at 10,000 ppm. The interaction between these two EOs also showed no interaction on the tested species. A combined formulation of oregano and thyme resulted in a synergistic effect, showing enhanced efficiency against A. flavus and A. parasiticus and P. chrysogenum. Mixtures of peppermint and tea tree produced synergistic effect against A. niger. Application of a modified Gompertz model considering fungal growth parameters like maximum colony diameter, maximum growth rate and lag time periods, under the various EO treatment scenarios, showed that the model could adequately describe and predict the growth of the tested fungi under these conditions.
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)