  • Search Results
  • High-Content Assay Multiplexing for Toxicity Screening in Induced Pluripotent Stem Cell-Derived Cardiomyocytes and Hepatocytes.
High-Content Assay Multiplexing for Toxicity Screening in Induced Pluripotent Stem Cell-Derived Cardiomyocytes and Hepatocytes.
Assay and drug development technologies (2015-11-06)
Fabian Alexander Grimm, Yasuhiro Iwata, Oksana Sirenko, Michael Bittner, Ivan Rusyn
ABSTRACT
Cell-based high-content screening (HCS) assays have become an increasingly attractive alternative to traditional in vitro and in vivo testing in pharmaceutical drug development and toxicological safety assessment. The time- and cost-effectiveness of HCS assays, combined with the organotypic nature of human induced pluripotent stem cell (iPSC)-derived cells, open new opportunities to employ physiologically relevant in vitro model systems to improve screening for potential chemical hazards. In this study, we used two human iPSC types, cardiomyocytes and hepatocytes, to test various high-content and molecular assay combinations for their applicability in a multiparametric screening format. Effects on cardiomyocyte beat frequency were characterized by calcium flux measurements for up to 90 min. Subsequent correlation with intracellular cAMP levels was used to determine if the effects on cardiac physiology were G-protein-coupled receptor dependent. In addition, we utilized high-content cell imaging to simultaneously determine cell viability, mitochondrial integrity, and reactive oxygen species (ROS) formation in both cell types. Kinetic analysis indicated that ROS formation is best detectable 30 min following initial treatment, whereas cytotoxic effects were most stable after 24 h. For hepatocytes, high-content imaging was also used to evaluate cytotoxicity and cytoskeletal integrity, as well as mitochondrial integrity and the potential for lipid accumulation. Lipid accumulation, a marker for hepatic steatosis, was most reliably detected 48 h following treatment with test compounds. Overall, our results demonstrate how a compendium of assays can be utilized for quantitative screening of chemical effects in iPSC cardiomyocytes and hepatocytes and enable rapid and cost-efficient multidimensional biological profiling of toxicity.
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Oncostatin M human, OSM, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Oncostatin M from mouse, BioReagent, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Oncostatin M human, BioReagent, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Tetraoctylammonium bromide, 98%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Cisapride monohydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Oncostatin-M from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %