  • Search Results
  • Magnetic nanoparticles with hydrophobicity and hydrophilicity for solid-phase extraction of polycyclic aromatic hydrocarbons from environmental water samples.
Magnetic nanoparticles with hydrophobicity and hydrophilicity for solid-phase extraction of polycyclic aromatic hydrocarbons from environmental water samples.
Journal of chromatography. A (2015-08-12)
Shu-Wen Xue, Min-Qiong Tang, Li Xu, Zhi-guo Shi
ABSTRACT
Magnetic nanoparticles (MNPs) featured with divinylbenzene (DVB) and sulfonate functionalities (Fe3O4-DVB-SO3(-)) were prepared via "thiol-ene" click chemistry. The hydrophobic DVB moieties were dedicated for extraction while the hydrophilic sulfonate groups were designed for dispersing the MNPs in aqueous sample solution. Thus, the specially designed material could ensure operational convenience and improve reproducibility during extraction. The application of the material was demonstrated by the extraction of polycyclic aromatic hydrocarbons (PAHs) from environmental water samples followed by gas chromatography-mass spectrometric analysis. The main factors influencing the extraction, including the type of the desorption solvent, the agitation mode, the amount of MNPs, extraction and desorption time and salt addition in sample solution, were investigated in detail. Under the optimized conditions, the proposed method showed satisfactory reproducibility with intra-day and inter-day relative standard deviations less than 16.5% and 21.2%, and low limits of detection of 1.1pgmL(-1), 0.8pgmL(-1), 1.1pgmL(-1), 1.4pgmL(-1), 0.6pgmL(-1), 2.1pgmL(-1) and 0.7pgmL(-1) for naphthalene, acenaphthene, fluorine, phenanthrene, anthracene, fluoranthene and pyrene, respectively. The developed method was also successfully used for determination of the PAHs in genuine lake and river environmental water samples by standard addition method. All the studied PAHs were detected in these waters with comparable results by the standard liquid-liquid extraction method. The developed MNPs with dual property of hydrophobicity and hydrophilicity were suitable for the treatment of water samples. The magnetic solid phase extraction based on this material was reliable and convenient. It has great potential in the preconcentration of trace analytes in complex matrix.
Product Description
SAFC
Acetic acid, glacial
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
Supelco
Ethyl acetate, HPLC grade, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG