  • Search Results
  • Optimized testing for C. trachomatis DNA in synovial fluid samples in clinical practice.
Optimized testing for C. trachomatis DNA in synovial fluid samples in clinical practice.
Zeitschrift fur Rheumatologie (2015-07-15)
J Freise, I Bernau, S Meier, H Zeidler, J G Kuipers
ABSTRACT
No standardized polymerase chain reaction (PCR) assay is available for detection of Chlamydia trachomatis (C. tr.) in synovial fluid (SF) for diagnostic use in clinical practice. This study tested the performance of two optimized molecular biology methods, to determine which is best suited for detecting C. tr. in SF clinical samples from patients with various rheumatologic diseases. Two DNA extraction methods, i.e., (1) alkaline lysis and (2) QIAEX II Gel Extraction Kit® + cetyltrimethylammonium bromide (CTAB; Qiagen, Hilden, Germany), and C. tr.-omp1-152 bp PCR were tested in SF samples from a total of 329 patients with the following diagnoses: reactive arthritis (ReA; n = 10, 4 patients had posturethritic ReA), undifferentiated arthritis (UA; n = 66), rheumatoid arthritis (RA; n = 169), psoriatic arthritis (PSA; n = 12), and osteoarthritis (OA) n = 72. In SF samples, C. tr.-omp1-152 bp PCR in combination with alkaline lysis DNA extraction allowed detection of more C. tr.-positive samples: 3/10 (30%) ReA patients (all with posturethritic ReA) and 20/66 (38%) UA patients were positive, compared to the 0/10 (0%) patients with ReA and 1/66 (2%) with UA detected using the QIAEX II Gel Extraction Kit® + CTAB. Moreover, 2/12 (17%) SF samples from PSA patients tested positive with alkaline lysis. All samples from patients with OA and RA tested negative. Alkaline lysis in combination with C. tr.-omp1-152 bp PCR emerged as the most sensitive method for identification of C. tr. in clinical SF samples.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis