  • Search Results
  • Conductive interpenetrating networks of polypyrrole and polycaprolactone encourage electrophysiological development of cardiac cells.
Conductive interpenetrating networks of polypyrrole and polycaprolactone encourage electrophysiological development of cardiac cells.
Acta biomaterialia (2015-09-27)
Benjamin S Spearman, Alexander J Hodge, John L Porter, John G Hardy, Zenda D Davis, Teng Xu, Xinyu Zhang, Christine E Schmidt, Michael C Hamilton, Elizabeth A Lipke
ABSTRACT
Conductive and electroactive polymers have the potential to enhance engineered cardiac tissue function. In this study, an interpenetrating network of the electrically-conductive polymer polypyrrole (PPy) was grown within a matrix of flexible polycaprolactone (PCL) and evaluated as a platform for directing the formation of functional cardiac cell sheets. PCL films were either treated with sodium hydroxide to render them more hydrophilic and enhance cell adhesion or rendered electroactive with PPy grown via chemical polymerization yielding PPy-PCL that had a resistivity of 1.0 ± 0.4 kΩ cm, which is similar to native cardiac tissue. Both PCL and PPy-PCL films supported cardiomyocyte attachment; increasing the duration of PCL pre-treatment with NaOH resulted in higher numbers of adherent cardiomyocytes per unit area, generating cell densities which were more similar to those on PPy-PCL films (1568 ± 126 cells mm(-2), 2880 ± 439 cells mm(-2), 3623 ± 456 cells mm(-2) for PCL with 0, 24, 48 h of NaOH pretreatment, respectively; 2434 ± 166 cells mm(-2) for PPy-PCL). When cardiomyocytes were cultured on the electrically-conductive PPy-PCL, more cells were observed to have peripheral localization of the gap junction protein connexin-43 (Cx43) as compared to cells on NaOH-treated PCL (60.3 ± 4.3% vs. 46.6 ± 5.7%). Cx43 gene expression remained unchanged between materials. Importantly, the velocity of calcium wave propagation was faster and calcium transient duration was shorter for cardiomyocyte monolayers on PPy-PCL (1612 ± 143 μm/s, 910 ± 63 ms) relative to cells on PCL (1129 ± 247 μm/s, 1130 ± 20 ms). In summary, PPy-PCL has demonstrated suitability as an electrically-conductive substrate for culture of cardiomyocytes, yielding enhanced functional properties; results encourage further development of conductive substrates for use in differentiation of stem cell-derived cardiomyocytes and cardiac tissue engineering applications. Current conductive materials for use in cardiac regeneration are limited by cytotoxicity or cost in implementation. In this manuscript, we demonstrate for the first time the application of a biocompatible, conductive polypyrrole-polycaprolactone film as a platform for culturing cardiomyocytes for cardiac regeneration. This study shows that the novel conductive film is capable of enhancing cell-cell communication through the formation of connexin-43, leading to higher velocities for calcium wave propagation and reduced calcium transient durations among cultured cardiomyocyte monolayers. Furthermore, it was demonstrated that chemical modification of polycaprolactone through alkaline-mediated hydrolysis increased overall cardiomyocyte adhesion. The results of this study provide insight into how cardiomyocytes interact with conductive substrates and will inform future research efforts to enhance the functional properties of cardiomyocytes, which is critical for their use in pharmaceutical testing and cell therapy.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.