  • Search Results
  • AZD2014, an Inhibitor of mTORC1 and mTORC2, Is Highly Effective in ER+ Breast Cancer When Administered Using Intermittent or Continuous Schedules.
AZD2014, an Inhibitor of mTORC1 and mTORC2, Is Highly Effective in ER+ Breast Cancer When Administered Using Intermittent or Continuous Schedules.
Molecular cancer therapeutics (2015-09-12)
Sylvie M Guichard, Jon Curwen, Teeru Bihani, Celina M D'Cruz, James W T Yates, Michael Grondine, Zoe Howard, Barry R Davies, Graham Bigley, Teresa Klinowska, Kurt G Pike, Martin Pass, Christine M Chresta, Urszula M Polanska, Robert McEwen, Oona Delpuech, Stephen Green, Sabina C Cosulich
ABSTRACT
mTOR is an atypical serine threonine kinase involved in regulating major cellular functions, such as nutrients sensing, growth, and proliferation. mTOR is part of the multiprotein complexes mTORC1 and mTORC2, which have been shown to play critical yet functionally distinct roles in the regulation of cellular processes. Current clinical mTOR inhibitors only inhibit the mTORC1 complex and are derivatives of the macrolide rapamycin (rapalogs). Encouraging effects have been observed with rapalogs in estrogen receptor-positive (ER(+)) breast cancer patients in combination with endocrine therapy, such as aromatase inhibitors. AZD2014 is a small-molecule ATP competitive inhibitor of mTOR that inhibits both mTORC1 and mTORC2 complexes and has a greater inhibitory function against mTORC1 than the clinically approved rapalogs. Here, we demonstrate that AZD2014 has broad antiproliferative effects across multiple cell lines, including ER(+) breast models with acquired resistance to hormonal therapy and cell lines with acquired resistance to rapalogs. In vivo, AZD2014 induces dose-dependent tumor growth inhibition in several xenograft and primary explant models. The antitumor activity of AZD2014 is associated with modulation of both mTORC1 and mTORC2 substrates, consistent with its mechanism of action. In combination with fulvestrant, AZD2014 induces tumor regressions when dosed continuously or using intermittent dosing schedules. The ability to dose AZD2014 intermittently, together with its ability to block signaling from both mTORC1 and mTORC2 complexes, makes this compound an ideal candidate for combining with endocrine therapies in the clinic. AZD2014 is currently in phase II clinical trials.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology