  • Search Results
  • A crystal structure of 2-hydroxybiphenyl 3-monooxygenase with bound substrate provides insights into the enzymatic mechanism.
A crystal structure of 2-hydroxybiphenyl 3-monooxygenase with bound substrate provides insights into the enzymatic mechanism.
Biochimica et biophysica acta (2015-08-16)
Margarita Kanteev, Almog Bregman-Cohen, Batel Deri, Anat Shahar, Noam Adir, Ayelet Fishman
ABSTRACT
2-Hydroxybiphenyl 3-monooxygenase (HbpA) is an FAD dependent monooxygenase which catalyzes the ortho-hydroxylation of a broad range of 2-substituted phenols in the presence of NADH and molecular oxygen. We have determined the structure of HbpA from the soil bacterium Pseudomonas azelaica HBP1 with bound 2-hydroxybiphenyl, as well as several variants, at a resolution of 2.3-2.5Å to investigate structure function correlations of the enzyme. An observed hydrogen bond between 2-hydroxybiphenyl and His48 in the active site confirmed the previously suggested role of this residue in substrate deprotonation. The entrance to the active site was confirmed by generating variant G255F which exhibited only 7% of the wild-type's specific activity of product formation, suggesting inhibition of substrate entrance into the active site by the large aromatic residue. Residue Arg242 is suggested to facilitate FAD movement and reduction as was previously reported in studies on the homologous protein para-hydroxybenzoate hydroxylase. In addition, it is suggested that Trp225, which is located in the active site, facilitates proper substrate entrance into the binding pocket in contrast to aklavinone-11-hydroxylase and para-hydroxybenzoate hydroxylase in which a residue at a similar position is responsible for substrate deprotonation. Structure function correlations described in this work will aid in the design of variants with improved activity and altered selectivity for potential industrial applications.
Sigma-Aldrich
2-Phenylphenol, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)