  • Search Results
  • Programmed packaging of mesoporous silica nanocarriers for matrix metalloprotease 2-triggered tumor targeting and release.
Programmed packaging of mesoporous silica nanocarriers for matrix metalloprotease 2-triggered tumor targeting and release.
Biomaterials (2015-05-06)
Zhen Zou, Xiaoxiao He, Dinggeng He, Kemin Wang, Zhihe Qing, Xue Yang, Li Wen, Jun Xiong, Liling Li, Linli Cai
ABSTRACT
The development of multifunctional nanocarrier with each unit functioning at the correct time and location is a challenge for clinical applications. With this in mind, a type of intelligent mesoporous silica nanocarrier (PGFMSN) is proposed for matrix metalloprotease 2 (MMP 2)-triggered tumor targeting and release by integrating programmed packing and MMP 2-degradable gelatin. Mesoporous silica nanoparticles (MSN) are first functionalized with folic acid (FA) as a target ligand to improve cell uptake. Then gelatin is introduced onto FA-MSN via temperature-induced gelation, where gelatin layer blocks drugs inside the mesopores and protects the targeting ligand. To prolong blood-circulation lifetime, PEG is further decorated to obtain PGFMSN. All units are programmatically incorporated in a simple way and coordinated in an optimal fashion. Cells, multicellular spheroids and in vivo results demonstrate that PGFMSN is shielded against nonspecific uptake. After circulating to tumor tissue, the up-regulated MMP-2 hydrolyzes gelatin layer to deshield PEG and switch on the function of FA, which facilitate the selective uptake by tumor cells through folate-receptor-mediated endocytosis. Meanwhile, the packaged drug is released due to the shedding of gelatin layer. It is shown that doxorubicin (DOX)-loaded exhibits superior tumor targeting, drug internalization, cytotoxicity, and antitumor efficacy over free DOX, non-PEGylated and non-targeted nanoparticles, which provides potential applications for targeted cancer therapy.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, BioReagent, suitable for fluorescence, ≥96.0% (HPCE)