  • Search Results
  • Selective X-ray contrast enhancement of the spleen of living mice mediated by gold nanorods.
Selective X-ray contrast enhancement of the spleen of living mice mediated by gold nanorods.
Contrast media & molecular imaging (2014-08-30)
Connor A Wathen, Chuck Caldwell, Nripen Chanda, Anandhi Upendran, Ajit Zambre, Zahra Afrasiabi, Sarah E Chapaman, Nathan Foje, W Matthew Leevy, Raghuraman Kannan
ABSTRACT
Gold nanomaterials (AuNPs) represent a promising new class of contrast agents for X-ray computed tomographic (CT) imaging in both research and clinical settings. These materials exhibit superior X-ray absorption properties compared with other iodinated agents, and thus require lower injection doses. Gold is nonimmunogenic and therefore contributes to safety profile in living specimens. Unfortunately, most reports on the use of AuNPs as X-ray CT enhancers only demonstrate marginal enhancement of the intended anatomical structure. In this study, we demonstrate the dramatic properties of gold nanorods (GNR) to serve as robust X-ray CT contrast-enhancing agent for selective imaging of the spleen. These organ-specific uptake properties were delineated by performing longitudinal CT imaging of living mice that were dosed with GNR at 2 day intervals. Rapid uptake in spleen was noted within 12 h of first systemic administration with a change in contrast enhancement of 90 Hounsfield units (ΔHU = 90) and with two subsequent injections a total contrast enhancement of over 200 HU was observed. The resulting images provide excellent contrast that will enable the detailed anatomical visualization and study of a range of pre-clinical models of spleen disease including infection and cancer.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Tantalum(V) ethoxide, packaged for use in deposition systems
SAFC
Hexadecyltrimethylammonium bromide
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%