  • Search Results
  • MicroRNA-29a Promotes Pancreatic Cancer Growth by Inhibiting Tristetraprolin.
MicroRNA-29a Promotes Pancreatic Cancer Growth by Inhibiting Tristetraprolin.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-09-12)
Xian-Jun Sun, Bing-Yan Liu, Shuo Yan, Ting-Hui Jiang, Hui-Qin Cheng, Hao-Sheng Jiang, Yan Cao, Ai-Wu Mao
ABSTRACT
The microRNA (miR) 29 family has been studied extensively for its involvement in several diseases, and aberrant expression of its members is associated with tumorigenesis and cancer progression. Here, we examined the role of miR-29a in pancreatic cancer and the involvement of tristetraprolin (TTP). We monitored miR-29a and TTP expression in pancreatic cancer by qRT-PCR and western blotting. The effect of miR-29a on pancreatic cancer was determined through MTT assay and migration assay. The results were validated in the tumorigenesis model. We found that miR-29a was up regulated in pancreatic tumor tissues and cell lines and positively correlated with metastasis. Ectopic expression of miR-29a increased the expression of pro-inflammatory factors and epithelial-mesenchymal transition (EMT) markers, through down regulating TTP. TTP was down regulated in tumor tissues, and its ectopic expression decreased cell viability and migration in vitro, inhibited tumor growth and the EMT phenotype in vivo, and reversed the effect of miR-29a on tumor cell proliferation and invasion in vitro and in vivo. Our results suggest that miR-29a acts as an oncogene by down regulating TTP and provide the basis for further studies exploring the potential of miR-29a and TTP as biomarkers and targets for the treatment of pancreatic cancer.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)