  • Search Results
  • Porphyromonas gingivalis RagB is a proinflammatory signal transducer and activator of transcription 4 agonist.
Porphyromonas gingivalis RagB is a proinflammatory signal transducer and activator of transcription 4 agonist.
Molecular oral microbiology (2014-11-25)
J A Hutcherson, J Bagaitkar, K Nagano, F Yoshimura, H Wang, D A Scott
ABSTRACT
Periodontal diseases are semi-ubiquitous and caused by chronic, plaque-induced inflammation. The 55-kDa immunodominant RagB outer membrane protein of Porphyromonas gingivalis, a keystone periodontal pathogen, has been proposed to facilitate nutrient transport. However, potential interactions between RagB and the innate response have not been examined. We determined that RagB exposure led to the differential and dose-related expression of multiple genes encoding proinflammatory mediators [interleukin-1α (IL-1α), IL-1β, IL-6, IL-8 and CCL2; all P < 0.05] in primary human monocytes and to the secretion of tumor necrosis factor and IL-8, but not interferon-γ or IL-12. RagB was shown to be a Toll-like receptor 2 (TLR2) and TLR4 agonist that activated signal transducer and activator of transcription 4 and nuclear factor-κB signaling, as determined by a combination of blocking antibodies, pharmaceutical inhibitors and gene silencing. In keeping, a ΔragB mutant similarly exhibited reduced inflammatory capacity, which was rescued by ragB complementation. These results suggest that RagB elicits a major pro-inflammatory response in primary human monocytes and, therefore, could play an important role in the etiology of periodontitis and systemic sequelae.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
D-(−)-Arabinose, ≥98%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Theobromine, ≥98.0%
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri