  • Search Results
  • Demethoxycurcumin was prior to temozolomide on inhibiting proliferation and induced apoptosis of glioblastoma stem cells.
Demethoxycurcumin was prior to temozolomide on inhibiting proliferation and induced apoptosis of glioblastoma stem cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-04-17)
Lei Shi, Xifeng Fei, Zhimin Wang
ABSTRACT
Temozolomide (TMZ) is widely used for treating glioblastoma (GBM), which can effectively inhibit the GBM growth for some months; however, it still could not prevent the invariable recurrence of GBM. The existence of glioma stem cells (GSCs) was considered to be a key factor. But TMZ has poor effects on GSCs. Recently, demethoxycurcumin (DMC) has been shown to display anti-tumor activities in malignant gliomas. However, its effects and the potential mechanisms on GSCs were still unclear. Our study showed that DMC was prior to TMZ on resulting in a significant increase in GSC apoptosis and a marked inhibition of cell growth in vitro. And combined treatment of DMC and TMZ showed more significant anti-GSC effects. Further research into the underlying mechanism demonstrated that this novel combinatorial regimen leads to changes of multiple cell signaling pathways including reactive oxygen species (ROS) production and caspase-3 signaling mitochondria-related apoptosis activation as well as inactivation of JAK/STAT3 signaling pathway. Taken together, our data demonstrate that the anti-GSC effects of DMC are better than TMZ, and combined treatment of DMC and TMZ has much stronger effects on GSCs.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O