  • Search Results
  • MicroRNA-153 suppresses the osteogenic differentiation of human mesenchymal stem cells by targeting bone morphogenetic protein receptor type II.
MicroRNA-153 suppresses the osteogenic differentiation of human mesenchymal stem cells by targeting bone morphogenetic protein receptor type II.
International journal of molecular medicine (2015-07-08)
Yujing Cao, Qiuxia LV, Cuitian LV
ABSTRACT
Elucidation of the molecular mechanisms governing the osteogenic differentiation of human mesenchymal stem cells (hMSCs) is of great importance for improving the treatment of bone-related diseases. MicroRNAs (miRNAs or miRs), a class of small non-coding RNAs, are critical in a number of biological processes, including the proliferation, differentiation and survival of cells and organisms. Emerging evidence indicates that miRNAs are essential in regulating osteoblastogenesis and bone formation. However, the role of miRNAs in osteoblast mechanotransduction remains to be defined. The present study aimed to examine the role of miR-153 in the osteogenesis of hMSCs and to investigate the impact of miR-153 on bone morphogenetic protein receptor type II (BMPR2) expression. The overexpression of miR-153 inhibited the osteogenic differentiation of hMSCs, whereas downregulation of miR-153 enhanced the process. Furthermore, bioinformatic analysis predicted that miR-153 is a potential regulator of BMPR2. The direct binding of miR-153 to the BMPR2 3'-untranslated region (3'-UTR) was demonstrated by a luciferase reporter assay using a construct containing the BMPR2 3'-UTR. In addition, knockdown of BMPR2 by RNA interference inhibited the osteogenic differentiation of hMSCs, with a similar effect to the upregulation of miR-153. In conclusion, the results suggest that miR-153 is a mechano-sensitive miRNA that regulates osteoblast differentiation by directly targeting BMPR2, and that therapeutic inhibition of miR-153 may be an efficient anabolic strategy for skeletal disorders caused by pathological mechanical loading.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis