  • Search Results
  • Repression of metadherin inhibits biological behavior of prostate cancer cells and enhances their sensitivity to cisplatin.
Repression of metadherin inhibits biological behavior of prostate cancer cells and enhances their sensitivity to cisplatin.
Molecular medicine reports (2015-02-17)
Yong-Bao Wei, Qiong Guo, Yun-Liang Gao, Bin Yan, Zhao Wang, Jin-Rui Yang, Wei Liu
ABSTRACT
Metadherin (MTDH), also known as astrocyte-elevated gene-1, was first cloned in 2002 and has been confirmed as an oncogene in numerous types of cancer by previous studies. Overexpression of MTDH has been observed in multiple types of cancer, including breast, esophageal, prostate, cervical and non-small-cell lung cancer, as well as neuroblastoma and hepatocellular carcinoma. However, at present, few investigations into MTDH‑associated prostate cancer have been performed. A previous study suggested that MTDH was expressed at higher levels in prostate cancer samples, compared with those of benign prostatic hyperplasia. The present study aimed to elucidate the effects of MTDH as an oncogene associated with the biological behavior of prostate cancer cells and chemotherapy-sensitivity to cisplatin in vitro. It was demonstrated that the inhibition of MTDH expression promoted cell apoptosis, reduced cell viability and weakened the invasive ability of prostate cancer cells. In addition, the suppression of MTDH expression increased cell sensitivity to cisplatin. Furthermore, it was demonstrated that MTDH‑associated phosphoinositide 3-kinase/Akt signaling pathways may be involved in mediating the biological behavior of prostate cancer.
Product Description
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles