MilliporeSigma
  • Response to infliximab therapy in ulcerative colitis is associated with decreased monocyte activation, reduced CCL2 expression and downregulation of Tenascin C.
Response to infliximab therapy in ulcerative colitis is associated with decreased monocyte activation, reduced CCL2 expression and downregulation of Tenascin C.
Journal of Crohn's & colitis (2014-12-18)
Maria K Magnusson, Hans Strid, Stefan Isaksson, Antal Bajor, Anders Lasson, Kjell-Arne Ung, Lena Öhman
ABSTRACT
The cellular mechanisms leading to infliximab therapy response in patients with ulcerative colitis (UC) are incompletely known. We therefore investigated early effects of infliximab therapy on monocytes and associated chemokines linked to clinical therapy response in UC patients. Blood and biopsies were obtained from anti-TNF therapy-naïve UC patients (n = 43) before (baseline) and during induction therapy with infliximab. Therapy response was evaluated at Week 14. Expression of monocyte activation markers and levels of chemokines in serum and biopsies were determined. Quantitative proteomic analysis was performed in cultured mucosal biopsies, and obtained data was validated in serum. In therapy responders, but not in non-responders, infliximab reduced blood monocyte expression of CD14 and CD86, 2 weeks after therapy commenced, relative to baseline. Serum CCL2 levels were decreased only among therapy responders at Week 2 and Week 14, relative to baseline. These data corresponded with lower levels of CD14, CD86 and CCL2 in intestinal tissue in responders as compared with non-responders at Week 14. Proteomic analysis of cultured biopsies showed that infliximab induced a reduction in Tenascin C that predicted downregulation of CCL2. Therapy responders, but not non-responders, had decreased serum Tenascin C levels at Week 2 and Week 14, relative to baseline. Infliximab therapy response in UC patients is associated with reduced monocyte activation and serum levels of CCL2 2 weeks after therapy commencement. In therapy responders, infliximab influenced Tenascin C, which might be a regulator of CCL2 expression and important for induction of the clinical therapy response.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)